2-Bromo-1-chloro-3,5-difluorobenzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC56086-5G
PC56086-25G
APOSPC56086-5GEA
4730
CZK
APOSPC56086-5G
APOSPC56086-25G
2-Bromo-1-chloro-3,5-difluorobenzene 95%
2-Bromo-1-chloro-3,5-difluorobenzene
Vzorec:
C₆H₂BrClF₂ MW: 227,434 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11849939 CAS číslo: 1020198-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...