3-(Trifluoromethylsulphonyl)phenylacetonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC56067-1G
PC56067-250MG
APOSPC56067-1GEA
1990
CZK
APOSPC56067-1G
APOSPC56067-250MG
3-(Trifluoromethylsulphonyl)phenylacetonitrile
3-(Trifluoromethylsulphonyl)phenylacetonitrile
Vzorec:
C₉H₆F₃NO₂S MW: 249,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29065960 CAS číslo: 1301739-15-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...