3,5-Dichloro-4-(trifluoromethyl)pyridin-2-ol
Dodavatel: Apollo Scientific
PC56049-1G
PC56049-5G
APOSPC56049-1GEA
7670
CZK
APOSPC56049-1G
APOSPC56049-5G
3,5-Dichloro-4-(trifluoromethyl)pyridin-2-ol
3,5-Dichloro-4-(trifluoromethyl)pyridin-2-ol
Vzorec:
C₆H₂Cl₂F₃NO MW: 231.987 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11227182 CAS číslo: 109919-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...