Tisk…

2-Nitro-5-(trifluoromethoxy)phenol

Dodavatel: Apollo Scientific
Varování

PC56046-1G PC56046-250MG
APOSPC56046-1GEA 12200 CZK
APOSPC56046-1G APOSPC56046-250MG
2-Nitro-5-(trifluoromethoxy)phenol
2-Nitro-5-(trifluoromethoxy)phenol
Vzorec: C₇H₄F₃NO₄
MW: 223.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527440
CAS číslo: 1197236-32-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR