Tisk…

3-(Trifluoromethylsulphonyl)benzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: [3-(Trifluoromethylsulphonyl)phenyl]methanol

PC56041-1G PC56041-250MG PC56041-5G
APOSPC56041-1GEA 34720 CZK
APOSPC56041-1G APOSPC56041-250MG APOSPC56041-5G
3-(Trifluoromethylsulphonyl)benzyl alcohol
3-(Trifluoromethylsulphonyl)benzyl alcohol
Vzorec: C₈H₇F₃O₃S
MW: 240,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25460315
CAS číslo: 1274903-38-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR