2,6-Difluoro-4-(trifluoromethyl)phenol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,5-Difluoro-4-hydroxybenzotrifluoride
PC56034-1G
PC56034-250MG
APOSPC56034-1GEA
3970
CZK
APOSPC56034-1G
APOSPC56034-250MG
2,6-Difluoro-4-(trifluoromethyl)phenol
2,6-Difluoro-4-(trifluoromethyl)phenol
Vzorec:
C₇H₃F₅O MW: 198,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09039179 CAS číslo: 276244-25-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...