5-Bromo-2-chloro-4-fluorobenzoic acid 94%
Dodavatel: Apollo Scientific
|
Varování
|
PC56027-1G
PC56027-250MG
PC56027-5G
APOSPC56027-1GEA
8500
CZK
APOSPC56027-1G
APOSPC56027-250MG
APOSPC56027-5G
5-Bromo-2-chloro-4-fluorobenzoic acid 94%
5-Bromo-2-chloro-4-fluorobenzoic acid
Vzorec:
C₇H₃BrClFO₂ MW: 253,453 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24107203 CAS číslo: 1204219-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...