4-Amino-3-bromo-2-fluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC56012-1G
PC56012-250MG
PC56012-5G
APOSPC56012-1GEA
4400
CZK
APOSPC56012-1G
APOSPC56012-250MG
APOSPC56012-5G
4-Amino-3-bromo-2-fluorobenzonitrile 95%
4-Amino-3-bromo-2-fluorobenzonitrile
Vzorec:
C₇H₄BrFN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD30297356 CAS číslo: 2090323-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...