Perfluoro(2,3-dimethylbutane) 80%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3-Bis(trifluoromethyl)-1,1,1,2,3,4,4,4-octafluorobutane
PC5567-25G
PC5567-5G
APOSPC5567-25GEA
13390
CZK
APOSPC5567-25G
APOSPC5567-5G
Perfluoro(2,3-dimethylbutane) 80%
Perfluoro(2,3-dimethylbutane)
Vzorec:
C₆F₁₄ Bod varu: 57…59 °C Hustota: 1,7729 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD07784184 CAS číslo: 354-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...