1-[2-Chloro-6-(trifluoromethyl)pyridin-3-yl]ethan-1-one 95%
Dodavatel: Apollo Scientific
PC55631-1G
PC55631-250MG
APOSPC55631-1GEA
23970
CZK
APOSPC55631-1G
APOSPC55631-250MG
1-[2-Chloro-6-(trifluoromethyl)pyridin-3-yl]ethan-1-one 95%
1-[2-Chloro-6-(trifluoromethyl)pyridin-3-yl]ethan-1-one
Vzorec:
C₈H₅ClF₃NO MW: 223,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10696225 CAS číslo: 944900-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...