2,6-Difluoro-4-(trifluoromethoxy)aniline 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC55523-1G
PC55523-250MG
PC55523-5G
APOSPC55523-1GEA
16370
CZK
APOSPC55523-1G
APOSPC55523-250MG
APOSPC55523-5G
2,6-Difluoro-4-(trifluoromethoxy)aniline 95%
2,6-Difluoro-4-(trifluoromethoxy)aniline
Vzorec:
C₇H₄F₅NO MW: 213,105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28790978 CAS číslo: 1100833-35-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...