3,6-Difluoro-2-iodopyridine
Dodavatel: Apollo Scientific
PC55436-1G
PC55436-250MG
APOSPC55436-1GEA
17990
CZK
APOSPC55436-1G
APOSPC55436-250MG
3,6-Difluoro-2-iodopyridine
3,6-Difluoro-2-iodopyridine
Vzorec:
C₅H₂F₂IN MW: 240,977 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185559 CAS číslo: 1214350-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...