Tisk…

3-(2,4-Difluorophenoxy)propanoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(2,4-Difluorophenoxy)propionic acid

PC5504-1G PC5504-5G
APOSPC5504-1GEA 2070 CZK
APOSPC5504-1G APOSPC5504-5G
3-(2,4-Difluorophenoxy)propanoic acid
3-(2,4-Difluorophenoxy)propanoic acid
Vzorec: C₉H₈F₂O₃
MW: 202,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764783
CAS číslo: 777-28-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR