Tisk…
3-(Trifluoromethyl)-1H-pyrazole-4-boronic acid, pinacol ester 98%
Dodavatel: Apollo Scientific
Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)-1H-pyrazole
PC5494-1G PC5494-250MG PC5494-5G
APOSPC5494-1GEA 6190 CZK
APOSPC5494-1G APOSPC5494-250MG APOSPC5494-5G
3-(Trifluoromethyl)-1H-pyrazole-4-boronic acid, pinacol ester 98%
3-(Trifluoromethyl)-1H-pyrazole-4-boronic acid, pinacol ester
Vzorec: C₁₀H₁₄BF₃N₂O₂
MW: 262.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501039
CAS číslo: 1218790-40-9