Tisk…

3,4-Bis(trifluoromethoxy)benzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: [3,4-Bis(trifluoromethoxy)phenyl]methanol

PC5478-250MG PC5478-1G
APOSPC5478-250MGEA 6620 CZK
APOSPC5478-250MG APOSPC5478-1G
3,4-Bis(trifluoromethoxy)benzyl alcohol 95%
3,4-Bis(trifluoromethoxy)benzyl alcohol
Vzorec: C₉H₆F₆O₃
MW: 276.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052963
CAS číslo: 1048922-46-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR