Tisk…

2'-(Difluoromethoxy)acetophenone 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-(Difluoromethoxy)phenyl)ethanone

PC5464-10G PC5464-5G PC5464-1G PC5464-25G
APOSPC5464-10GEA 10620 CZK
APOSPC5464-10G APOSPC5464-5G APOSPC5464-1G APOSPC5464-25G
2'-(Difluoromethoxy)acetophenone 98%
2'-(Difluoromethoxy)acetophenone
Vzorec: C₉H₈F₂O₂
MW: 186,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236219
CAS číslo: 127842-55-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR