2-Iodo-1-(trifluoromethoxy)tetrafluoroethane 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Iodotetrafluoroethyl trifluoromethyl ether
PC5449-1G
PC5449-25G
PC5449-5G
APOSPC5449-1GEA
3310
CZK
APOSPC5449-1G
APOSPC5449-25G
APOSPC5449-5G
2-Iodo-1-(trifluoromethoxy)tetrafluoroethane 97%
2-Iodo-1-(trifluoromethoxy)tetrafluoroethane
Vzorec:
C₃F₇IO MW: 311.925 g/mol Bod varu: 47.5 °C Hustota: 2.086 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00155969 CAS číslo: 1561-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...