Perfluoro(tetrahydrofuran) 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Octafluorotetrahydrofuran
PC5424-100G
PC5424-25G
APOSPC5424-100GEA
35710
CZK
APOSPC5424-100G
APOSPC5424-25G
Perfluoro(tetrahydrofuran) 99%
Perfluoro(tetrahydrofuran)
Vzorec:
C₄F₈O Bod varu: –0,8 °C Bod tání: –85 °C Hustota: 1,52 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00465561 CAS číslo: 773-14-8 UN: 3163 ADR: 2.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...