Tisk…

Ethyl 3-bromo-2,2-difluoropropionate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 3-bromo-2,2-difluoropropanoate

PC540091-1G PC540091-5G
APOSPC540091-1GEA 1910 CZK
APOSPC540091-1G APOSPC540091-5G
Ethyl 3-bromo-2,2-difluoropropionate 95%
Ethyl 3-bromo-2,2-difluoropropionate
Vzorec: C₅H₇BrF₂O₂
MW: 217,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144891
CAS číslo: 111773-24-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR