Tisk…

2-Fluoropropenoic acid butyl ester 95%

Dodavatel: Apollo Scientific

PC540090-100MG PC540090-1G PC540090-250MG
APOSPC540090-100MGEA 2860 CZK
APOSPC540090-100MG APOSPC540090-1G APOSPC540090-250MG
2-Fluoropropenoic acid butyl ester 95%
2-Fluoropropenoic acid butyl ester
Vzorec: C₇H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD00190087
CAS číslo: 10011-39-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR