Tisk…

2,3,4,5-Tetrafluoro-6-iodobenzoic acid ≥98%

Dodavatel: Apollo Scientific

PC540045-250MG PC540045-500MG
APOSPC540045-250MGEA 4700 CZK
APOSPC540045-250MG APOSPC540045-500MG
2,3,4,5-Tetrafluoro-6-iodobenzoic acid ≥98%
2,3,4,5-Tetrafluoro-6-iodobenzoic acid
Vzorec: C₇HF₄IO₂
MW: 319,98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28359658
CAS číslo: 110625-15-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR