1-Bromo-2-(bromomethyl)-3,4,5-trifluorobenzene 95%
Dodavatel: Apollo Scientific
PC53845-1G
PC53845-250MG
PC53845-5G
APOSPC53845-1GEA
14880
CZK
APOSPC53845-1G
APOSPC53845-250MG
APOSPC53845-5G
1-Bromo-2-(bromomethyl)-3,4,5-trifluorobenzene 95%
1-Bromo-2-(bromomethyl)-3,4,5-trifluorobenzene
Vzorec:
C₇H₃Br₂F₃ MW: 303,902 g/mol Storage Temperature: Ambient |
CAS číslo:
1936711-52-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...