Tisk…

Methyl 1-(difluoromethyl)pyrazole-4-carboxylate 95%

Dodavatel: Apollo Scientific

PC53601-1G PC53601-250MG
APOSPC53601-1GEA 18620 CZK
APOSPC53601-1G APOSPC53601-250MG
Methyl 1-(difluoromethyl)pyrazole-4-carboxylate 95%
Methyl 1-(difluoromethyl)pyrazole-4-carboxylate
Vzorec: C₆H₆F₂N₂O₂
MW: 176,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31417710
CAS číslo: 2101196-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR