Tisk…

Methyl 3,5-dichloro-4-fluoro-benzoate 95%

Dodavatel: Apollo Scientific

PC53598-1G PC53598-250MG
APOSPC53598-1GEA 11810 CZK
APOSPC53598-1G APOSPC53598-250MG
Methyl 3,5-dichloro-4-fluoro-benzoate 95%
Methyl 3,5-dichloro-4-fluoro-benzoate
Vzorec: C₈H₅Cl₂FO₂
MW: 223.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406866
CAS číslo: 1214375-18-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR