Tisk…

2-Chloro-4-(trifluoromethoxy)pyridine 97%

Dodavatel: Apollo Scientific
Varování

PC53587-100MG PC53587-250MG
APOSPC53587-100MGEA 3410 CZK
APOSPC53587-100MG APOSPC53587-250MG
2-Chloro-4-(trifluoromethoxy)pyridine 97%
2-Chloro-4-(trifluoromethoxy)pyridine
Vzorec: C₆H₃ClF₃NO
MW: 197,542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20492606
CAS číslo: 1206975-33-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR