Tisk…

1,2-Dibromo-3-chloro-5-fluorobenzene 96%

Dodavatel: Apollo Scientific
Varování

PC53579-1G PC53579-5G PC53579-25G
APOSPC53579-1GEA 1240 CZK
APOSPC53579-1G APOSPC53579-5G APOSPC53579-25G
1,2-Dibromo-3-chloro-5-fluorobenzene 96%
1,2-Dibromo-3-chloro-5-fluorobenzene
Vzorec: C₆H₂Br₂ClF
MW: 288,339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878108
CAS číslo: 1000577-62-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR