1-Bromo-2-iodo-3-nitro-5-(trifluoromethoxy)benzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC53577-1G
PC53577-250MG
PC53577-5G
APOSPC53577-1GEA
16340
CZK
APOSPC53577-1G
APOSPC53577-250MG
APOSPC53577-5G
1-Bromo-2-iodo-3-nitro-5-(trifluoromethoxy)benzene 95%
1-Bromo-2-iodo-3-nitro-5-(trifluoromethoxy)benzene
Vzorec:
C₇H₂BrF₃INO₃ MW: 411,899 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31710613 CAS číslo: 2366994-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...