3-Chloro-4-fluoro-5-(trifluoromethyl)benzaldehyde 97%
Dodavatel: Apollo Scientific
PC53573-1G
PC53573-250MG
PC53573-5G
APOSPC53573-1GEA
14880
CZK
APOSPC53573-1G
APOSPC53573-250MG
APOSPC53573-5G
3-Chloro-4-fluoro-5-(trifluoromethyl)benzaldehyde 97%
3-Chloro-4-fluoro-5-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₃ClF₄O MW: 226,555 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13192651 CAS číslo: 134099-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...