Tisk…

2,4-Difluoro-DL-phenylglycine 98%

Dodavatel: Apollo Scientific
Varování

PC5357-1G PC5357-250MG PC5357-5G
APOSPC5357-1GEA 1450 CZK
APOSPC5357-1G APOSPC5357-250MG APOSPC5357-5G
2,4-Difluoro-DL-phenylglycine 98%
2,4-Difluoro-DL-phenylglycine
Vzorec: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD02662503
CAS číslo: 240409-02-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR