Tisk…

2-Amino-3-bromo-4,5-difluorobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC53568-1G PC53568-250MG PC53568-5G
APOSPC53568-1GEA 1040 CZK
APOSPC53568-1G APOSPC53568-250MG APOSPC53568-5G
2-Amino-3-bromo-4,5-difluorobenzoic acid 95%
2-Amino-3-bromo-4,5-difluorobenzoic acid
Vzorec: C₇H₄BrF₂NO₂
MW: 252,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06808541
CAS číslo: 874831-41-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR