Tisk…

2-[(2-Fluorophenyl)methylsulphanyl]acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-((2-Fluorobenzyl)thio)acetic acid

PC53534-250MG PC53534-100MG
APOSPC53534-250MGEA 0 CZK
APOSPC53534-250MG APOSPC53534-100MG
2-[(2-Fluorophenyl)methylsulphanyl]acetic acid
2-[(2-Fluorophenyl)methylsulphanyl]acetic acid
Vzorec: C₉H₉FO₂S
MW: 200,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09809971
CAS číslo: 180044-55-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR