Tisk…

2-[(2-Chloro-6-fluorophenyl)methylsulphanyl]acetic acid

Dodavatel: Apollo Scientific
Varování

PC53532-1G PC53532-250MG PC53532-100MG
APOSPC53532-1GEA 13260 CZK
APOSPC53532-1G APOSPC53532-250MG APOSPC53532-100MG
2-[(2-Chloro-6-fluorophenyl)methylsulphanyl]acetic acid
2-[(2-Chloro-6-fluorophenyl)methylsulphanyl]acetic acid
Vzorec: C₉H₈ClFO₂S
MW: 234,675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05815052
CAS číslo: 714260-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR