Tisk…

4-(Carbamoylmethoxy)-3-fluorobenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC53531-1G PC53531-250MG PC53531-100MG
APOSPC53531-1GEA 18650 CZK
APOSPC53531-1G APOSPC53531-250MG APOSPC53531-100MG
4-(Carbamoylmethoxy)-3-fluorobenzenesulphonyl chloride
4-(Carbamoylmethoxy)-3-fluorobenzenesulphonyl chloride
Vzorec: C₈H₇ClFNO₄S
MW: 267,662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24328298
CAS číslo: 1443292-54-3
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR