Tisk…

4-Chloro-7-fluoro-1H-imidazo[4,5-c]pyridine 95+%

Dodavatel: Apollo Scientific
Varování

PC535086-100MG PC535086-250MG
APOSPC535086-100MGEA 2480 CZK
APOSPC535086-100MG APOSPC535086-250MG
4-Chloro-7-fluoro-1H-imidazo[4,5-c]pyridine 95+%
4-Chloro-7-fluoro-1H-imidazo[4,5-c]pyridine
Vzorec: C₆H₃ClFN₃
MW: 171,56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18253397
CAS číslo: 405230-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR