Tisk…

Methyl 2-fluoro-4-(methylsulphonyl)benzoate

Dodavatel: Apollo Scientific
Varování

PC53508-1G PC53508-250MG PC53508-5G
APOSPC53508-1GEA 4960 CZK
APOSPC53508-1G APOSPC53508-250MG APOSPC53508-5G
Methyl 2-fluoro-4-(methylsulphonyl)benzoate
Methyl 2-fluoro-4-(methylsulphonyl)benzoate
Vzorec: C₉H₉FO₄S
Storage Temperature: Ambient
MDL Number: MFCD27578943
CAS číslo: 1820716-37-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR