Tisk…

5-Fluorobenzofuran-6-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

PC535078-1G PC535078-250MG
APOSPC535078-1GEA 4200 CZK
APOSPC535078-1G APOSPC535078-250MG
5-Fluorobenzofuran-6-carboxylic acid 97%
5-Fluorobenzofuran-6-carboxylic acid
Vzorec: C₉H₅FO₃
MW: 180,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20661151
CAS číslo: 1312556-72-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR