(5-Fluorobenzofuran-2-yl)boronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC535073-100MG
PC535073-250MG
APOSPC535073-100MGEA
1820
CZK
APOSPC535073-100MG
APOSPC535073-250MG
(5-Fluorobenzofuran-2-yl)boronic acid 98%
(5-Fluorobenzofuran-2-yl)boronic acid
Vzorec:
C₈H₆BFO₃ MW: 179,941 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD18383019 CAS číslo: 473416-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...