Tisk…

8-Bromo-6-fluoro-2-methylquinazolin-4(3H)-one 95+%

Dodavatel: Apollo Scientific
Varování

PC535071-250MG PC535071-1G
APOSPC535071-250MGEA 1330 CZK
APOSPC535071-250MG APOSPC535071-1G
8-Bromo-6-fluoro-2-methylquinazolin-4(3H)-one 95+%
8-Bromo-6-fluoro-2-methylquinazolin-4(3H)-one
Vzorec: C₉H₆BrFN₂O
MW: 257,059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27996279
CAS číslo: 1352717-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR