Tisk…

Methyl 4-fluorothiophene-2-carboxylate 95+%

Dodavatel: Apollo Scientific
Varování

PC535066-250MG PC535066-1G
APOSPC535066-250MGEA 2860 CZK
APOSPC535066-250MG APOSPC535066-1G
Methyl 4-fluorothiophene-2-carboxylate 95+%
Methyl 4-fluorothiophene-2-carboxylate
Vzorec: C₆H₅FO₂S
MW: 160,166 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11618006
CAS číslo: 32431-75-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR