Tisk…

5-Chloro-4-fluoropyridin-2-amine 97%

Dodavatel: Apollo Scientific
Varování

PC535063-1G PC535063-250MG
APOSPC535063-1GEA 4600 CZK
APOSPC535063-1G APOSPC535063-250MG
5-Chloro-4-fluoropyridin-2-amine 97%
5-Chloro-4-fluoropyridin-2-amine
Vzorec: C₅H₄ClFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD22547000
CAS číslo: 1393574-54-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR