Tisk…

2,4-Dichloro-6-fluoropyrimidine 95%

Dodavatel: Apollo Scientific
Varování

PC535062-100MG PC535062-250MG
APOSPC535062-100MGEA 5960 CZK
APOSPC535062-100MG APOSPC535062-250MG
2,4-Dichloro-6-fluoropyrimidine 95%
2,4-Dichloro-6-fluoropyrimidine
Vzorec: C₄HCl₂FN₂
MW: 166,969 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16619134
CAS číslo: 3833-57-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR