Tisk…

3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%

Dodavatel: Apollo Scientific
Varování

PC535059-100MG PC535059-250MG
APOSPC535059-100MGEA 7610 CZK
APOSPC535059-100MG APOSPC535059-250MG
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide
Vzorec: C₇H₅BrF₃N·BrH
Storage Temperature: Refrigerator
MDL Number: MFCD29760335
CAS číslo: 1384972-85-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR