2,2,2-Trifluoro-1-(3-fluoropyridin-2-yl)ethanone 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC535057-100MG
PC535057-250MG
APOSPC535057-100MGEA
1490
CZK
APOSPC535057-100MG
APOSPC535057-250MG
2,2,2-Trifluoro-1-(3-fluoropyridin-2-yl)ethanone 95%
2,2,2-Trifluoro-1-(3-fluoropyridin-2-yl)ethanone
Vzorec:
C₇H₃F₄NO Storage Temperature: Ambient |
MDL Number:
MFCD13189065 CAS číslo: 1060802-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...