3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC53505-1G
PC53505-250MG
PC53505-5G
APOSPC53505-1GEA
5100
CZK
APOSPC53505-1G
APOSPC53505-250MG
APOSPC53505-5G
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Vzorec:
C₇H₈BFO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD19687060 CAS číslo: 1146614-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...