6-Bromo-3-fluoropyridin-2-amine 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC535049-100MG
PC535049-250MG
APOSPC535049-100MGEA
1580
CZK
APOSPC535049-100MG
APOSPC535049-250MG
6-Bromo-3-fluoropyridin-2-amine 98%
6-Bromo-3-fluoropyridin-2-amine
Vzorec:
C₅H₄BrFN₂ MW: 191,001 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28368857 CAS číslo: 1379457-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...