2-Bromo-6-fluoro-3-methylpyridine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC535035-100MG
PC535035-250MG
APOSPC535035-100MGEA
6580
CZK
APOSPC535035-100MG
APOSPC535035-250MG
2-Bromo-6-fluoro-3-methylpyridine 95+%
2-Bromo-6-fluoro-3-methylpyridine
Vzorec:
C₆H₅BrFN MW: 190,013 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16988804 CAS číslo: 1211536-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...