Tisk…

4-Bromo-3-fluoro-2-methylpyridine 95+%

Dodavatel: Apollo Scientific
Varování

PC535034-100MG PC535034-250MG
APOSPC535034-100MGEA 3970 CZK
APOSPC535034-100MG APOSPC535034-250MG
4-Bromo-3-fluoro-2-methylpyridine 95+%
4-Bromo-3-fluoro-2-methylpyridine
Vzorec: C₆H₅BrFN
MW: 190.013 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16609967
CAS číslo: 1211583-78-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR