Tisk…

5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one 95+%

Dodavatel: Apollo Scientific
Varování

PC535030-1G PC535030-250MG
APOSPC535030-1GEA 4370 CZK
APOSPC535030-1G APOSPC535030-250MG
5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one 95+%
5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one
Vzorec: C₅H₂BrF₃N₂O
MW: 242,981 g/mol
Storage Temperature: Freezer
MDL Number: MFCD15528725
CAS číslo: 785777-90-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR