2-Bromo-3-fluorobenzal fluoride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-1-(difluoromethyl)-3-fluorobenzene
PC53503-1G
PC53503-5G
PC53503-250MG
APOSPC53503-1GEA
7440
CZK
APOSPC53503-1G
APOSPC53503-5G
APOSPC53503-250MG
2-Bromo-3-fluorobenzal fluoride
2-Bromo-3-fluorobenzal fluoride
Vzorec:
C₇H₄BrF₃ MW: 225.006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14698531 CAS číslo: 1214338-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...