Tisk…

3-Chloro-2,4-difluoropyridine 95+%

Dodavatel: Apollo Scientific
Varování

PC535029-1G PC535029-250MG
APOSPC535029-1GEA 6090 CZK
APOSPC535029-1G APOSPC535029-250MG
3-Chloro-2,4-difluoropyridine 95+%
3-Chloro-2,4-difluoropyridine
Vzorec: C₅H₂ClF₂N
MW: 149,526 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09998888
CAS číslo: 851179-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR